Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAP4K2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001307990.1 | NP_001294919.1 | 22 | 1605 | ||
NM_004579.4 | NP_004570.2 | 22 | 1629 | |||
XM_011545203.2 | XP_011543505.1 | 22 | 1623 | |||
XM_011545204.2 | XP_011543506.1 | 21 | 1491 | |||
XM_017018093.1 | XP_016873582.1 | 21 | 1467 | |||
XM_017018094.1 | XP_016873583.1 | 21 | 1523 | |||
XM_017018095.1 | XP_016873584.1 | 21 | 1523 | |||
XM_017018096.1 | XP_016873585.1 | 22 | 1623 | |||
GenBank mRNA | AK298383.1 | - | 22 | 1566 | ||
BC039839.1 | - | 22 | 1512 | |||
BC047865.1 | - | 22 | 1592 | 5530523 | ||
U07349.1 | - | 22 | 1560 |