Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHUK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278.4 | NP_001269.3 | 13 | 1545 | ||
NM_001320928.1 | NP_001307857.1 | 13 | 1545 | |||
XM_017015611.1 | XP_016871100.1 | 13 | 1545 | |||
XM_017015612.1 | XP_016871101.1 | 13 | 1545 | |||
XM_017015613.1 | XP_016871102.1 | 3 | 360 | |||
GenBank mRNA | AB209028.1 | - | 13 | 1499 | ||
AF009225.1 | - | 13 | 1502 | |||
AF012890.1 | - | 13 | 1467 | |||
AF080157.1 | - | 13 | 1562 | |||
BC092514.1 | - | 13 | 1483 | 3063613 | ||
BM800404.1 | - | 730 | ||||
U22512.1 | - | 12 | 1375 |