Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAP3K12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001193511.1 | NP_001180440.1 | 7 | 1466 | ||
NM_006301.3 | NP_006292.3 | 8 | 1367 | |||
XM_005269138.3 | XP_005269195.1 | 7 | 1394 | |||
XM_005269140.2 | XP_005269197.1 | 7 | 1307 | |||
XM_006719588.3 | XP_006719651.1 | 7 | 1288 | |||
XM_011538725.2 | XP_011537027.1 | 7 | 2271 | |||
XM_017019956.1 | XP_016875445.1 | 8 | 1295 | |||
GenBank mRNA | AB209453.1 | - | 2 | 2317 | ||
AK094195.1 | - | 7 | 1262 | |||
AK314530.1 | - | 7 | 1614 | |||
BC037585.1 | - | 4 | 376 | |||
BC050050.1 | - | 7 | 1467 | 4824512 | ||
U07358.1 | - | 8 | 1194 |