Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NFATC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004555.3 | NP_004546.1 | 9 | 2787 | ||
NM_173163.2 | NP_775186.1 | 9 | 2787 | |||
NM_173165.2 | NP_775188.1 | 9 | 2787 | |||
GenBank mRNA | AB209824.1 | - | 7 | 1414 | ||
AK290370.1 | - | 9 | 2527 | |||
BC001050.2 | - | 9 | 2505 | 3505967 | ||
EU887605.1 | - | 9 | 2471 | |||
EU887606.1 | - | 9 | 2471 | |||
EU887607.1 | - | 9 | 2471 | |||
EU887608.1 | - | 9 | 2798 | |||
EU887609.1 | - | 9 | 2798 | |||
EU887610.1 | - | 9 | 2798 | |||
EU887611.1 | - | 9 | 2516 | |||
EU887612.1 | - | 9 | 2516 | |||
EU887613.1 | - | 9 | 2516 | |||
EU887614.1 | - | 9 | 2424 | |||
EU887615.1 | - | 9 | 2424 | |||
EU887616.1 | - | 9 | 2424 | |||
EU887617.1 | - | 9 | 2429 | |||
EU887618.1 | - | 9 | 2429 | |||
EU887619.1 | - | 9 | 2429 | |||
EU887620.1 | - | 9 | 2466 | |||
EU887621.1 | - | 9 | 2466 | |||
EU887622.1 | - | 9 | 2466 | |||
EU887623.1 | - | 9 | 2870 | |||
EU887624.1 | - | 9 | 2870 | |||
EU887625.1 | - | 9 | 2870 | |||
L41067.1 | - | 9 | 2681 | |||
U14510.1 | - | 9 | 2495 | |||
U85428.1 | - | 9 | 2495 | |||
U85429.1 | - | 9 | 2495 | |||
U85430.1 | - | 9 | 2495 |