Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TCL1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098725.1 | NP_001092195.1 | 1, 2 | 241 | ||
NM_021966.2 | NP_068801.1 | 1, 2 | 241 | |||
XM_017021676.1 | XP_016877165.1 | 1, 2 | 197 | |||
XM_017021677.1 | XP_016877166.1 | 1, 2 | 197 | |||
GenBank mRNA | BC003574.1 | - | 1, 2 | 153 | 3544215 | |
BC005831.2 | - | 1, 2 | 153 | 2966563 | ||
BC014024.1 | - | 1, 2 | 157 | 4308655 | ||
BG718395.1 | - | 1, 2 | 246 | |||
CR456796.1 | - | 1, 2 | 111 | |||
X82240.1 | - | 1, 2 | 156 |