Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: JAK3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000215.3 | NP_000206.2 | 12 | 1693 | ||
XM_005259896.3 | XP_005259953.1 | 12 | 1732 | |||
XM_011527990.1 | XP_011526292.1 | 12 | 1732 | |||
XM_011527991.2 | XP_011526293.1 | 12 | 1732 | |||
GenBank mRNA | BC028068.1 | - | 12 | 1683 | 5212575 | |
KU178153.1 | - | 11 | 1593 | |||
KU178154.1 | - | 12 | 1707 | |||
U09607.1 | - | 11 | 1688 | |||
U31601.1 | - | 12 | 1688 |