Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: JAK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320923.1 | NP_001307852.1 | 22 | 3115 | ||
NM_001321852.1 | NP_001308781.1 | 21 | 3118 | |||
NM_001321853.1 | NP_001308782.1 | 23 | 3377 | |||
NM_001321854.1 | NP_001308783.1 | 22 | 3293 | |||
NM_001321855.1 | NP_001308784.1 | 22 | 3296 | |||
NM_001321856.1 | NP_001308785.1 | 21 | 3031 | |||
NM_001321857.1 | NP_001308786.1 | 21 | 3209 | |||
NM_002227.3 | NP_002218.2 | 21 | 3212 | |||
XM_017001212.1 | XP_016856701.1 | 21 | 3056 | |||
GenBank mRNA | AB209057.1 | - | 20 | 2968 | ||
AB219242.1 | - | 19 | 2859 | |||
AK314257.1 | - | 22 | 3116 | |||
BC132729.1 | - | 20 | 2930 | 40146751 | ||
BX647675.1 | - | 7 | 816 | |||
HQ628631.1 | - | 19 | 2892 | |||
M64174.1 | - | 20 | 2937 |