Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STAT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005419.3 | NP_005410.1 | 2 | 219 | ||
NM_198332.1 | NP_938146.1 | 2 | 219 | |||
XM_005269111.3 | XP_005269168.2 | 2 | 127 | |||
XM_011538697.2 | XP_011536999.1 | 2 | 127 | |||
XM_011538698.2 | XP_011537000.1 | 2 | 100 | |||
XM_011538699.2 | XP_011537001.1 | 2 | 127 | |||
XM_017019904.1 | XP_016875393.1 | 2 | 127 | |||
GenBank mRNA | AK295052.1 | - | 2 | 107 | ||
AK295114.1 | - | 2 | 121 | |||
AK296939.1 | - | 2 | 92 | |||
AK296940.1 | - | 2 | 92 | |||
AK308051.1 | - | 2 | 97 | |||
AK310727.1 | - | 2 | 73 | |||
AK316516.1 | - | 2 | 103 | |||
BC006092.1 | - | 1 | 18 | |||
BC051284.1 | - | 1 | 39 | 6460486 | ||
CD684210.1 | - | 2 | 115 | |||
DB482283.1 | - | 2 | 219 | |||
M97934.1 | - | 2 | 53 |