Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STAT6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001178078.1 | NP_001171549.1 | 13 | 1678 | ||
NM_001178079.1 | NP_001171550.1 | 13 | 1604 | |||
NM_001178080.1 | NP_001171551.1 | 12 | 1522 | |||
NM_001178081.1 | NP_001171552.1 | 11 | 1383 | |||
NM_003153.4 | NP_003144.3 | 13 | 1659 | |||
NR_033659.1 | 12 | 1520 | ||||
XM_006719575.2 | XP_006719638.1 | 14 | 1656 | |||
XM_011538703.2 | XP_011537005.1 | 13 | 1679 | |||
XM_011538704.2 | XP_011537006.1 | 14 | 1690 | |||
XM_011538705.2 | XP_011537007.1 | 14 | 1800 | |||
XM_011538706.2 | XP_011537008.1 | 14 | 1656 | |||
XM_011538707.2 | XP_011537009.1 | 13 | 1641 | |||
XM_011538708.2 | XP_011537010.1 | 12 | 1483 | |||
XM_011538709.2 | XP_011537011.1 | 13 | 1517 | |||
GenBank mRNA | AB103089.1 | - | 7 | 831 | ||
AK290431.1 | - | 13 | 1607 | |||
AK291044.1 | - | 13 | 1605 | |||
AK298721.1 | - | 11 | 1203 | |||
AK299094.1 | - | 12 | 1456 | |||
AK308751.1 | - | 13 | 1499 | |||
AK312471.1 | - | 13 | 1525 | |||
AK316142.1 | - | 12 | 1466 | |||
AK316236.1 | - | 12 | 1297 | |||
BC075852.1 | - | 13 | 1460 | 5530399 | ||
BT007380.1 | - | 12 | 1353 | |||
U16031.1 | - | 13 | 1518 |