Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atp2a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027839.1 | 20, 21 | 4888 | |||
GenBank mRNA | CO404645.1 | - | 1, 2 | 527 | ||
FQ213049.1 | - | 1, 2 | 544 | |||
J04022.1 | - | 20, 21 | 4886 |