Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atp2a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027839.1 | 20 | 4501 | |||
GenBank mRNA | CO404645.1 | - | 1 | 140 | ||
FQ213049.1 | - | 1 | 157 | |||
J04022.1 | - | 20 | 4499 | |||
J04024.1 | - | 20 | 4233 |