Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 4931440P22Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027955.1 | 2 | 351 | |||
GenBank mRNA | AK136269.1 | - | 2 | 366 | ||
AK170145.1 | - | 2 | 317 | |||
AK171113.1 | - | 1 | 222 |