Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 4930483J18Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015603.1 | 2 | 812 | |||
GenBank mRNA | AK015615.1 | - | 2 | 692 | ||
AK016854.1 | - | 2 | 813 | |||
BC049560.1 | - | 4 | 735 | |||
BC087915.1 | - | 1 | 410 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY716529.1 | - | 2 | 813 | Chr. 15 - 81190853 - 81192603 [+] on Build GRCm38 |