Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 2410018L13Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015504.1 | 4 | 1008 | |||
GenBank mRNA | AK134640.1 | - | 4 | 1009 | ||
BC063063.1 | - | 5 | 2978 |
Gene Symbol: Gm9222 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC031503.1 | - | 1527 |