Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 4932414J04Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_028259.1 | 2 | 125 | |||
GenBank mRNA | AK030001.1 | - | 2 | 126 |
Gene Symbol: 4932415M13Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_073205.1 | 2 | 98 | |||
GenBank mRNA | AK030017.1 | - | 2 | 99 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB616445.1 | - | 2 | 99 | Chr. 17 - 53716509 - 53724143 [+] on Build GRCm38 |