Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cep83os | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015524.1 | 3 | 2321 | |||
GenBank mRNA | AK016525.1 | - | 3 | 2281 | ||
AK144701.1 | - | 3 | 2284 | |||
AK154718.1 | - | 3 | 2254 | |||
AK155868.1 | - | 3 | 2254 | |||
BC021358.1 | - | 1 | 1099 | |||
BC061482.1 | - | 1 | 1297 | |||
BC067070.1 | - | 3 | 2267 |