Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mir99ahg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015543.2 | 4 | 609 | |||
GenBank mRNA | AK012955.1 | - | 482 | |||
AK021336.1 | - | 653 | ||||
AK033222.1 | - | 214 | ||||
AK048031.1 | - | 664 | ||||
AK083771.1 | - | 4 | 610 | |||
AK144299.1 | - | 629 | ||||
AK148800.1 | - | 550 |