Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 6530411M01Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027881.1 | 1 | 518 | |||
GenBank mRNA | AK020377.1 | - | 1 | 519 | ||
AK035915.1 | - | 1 | 421 | |||
AK078247.1 | - | 1 | 333 | |||
BY721399.1 | - | 1 | 519 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY718979.1 | - | 1 | 333 | Chr. 17 - 9149174 - 9167836 [-] on Build GRCm38 |