Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 1700029M20Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015613.1 | 3 | 1040 | |||
NR_027909.1 | 2 | 911 | ||||
GenBank mRNA | AK006510.2 | - | 3 | 925 | ||
AK015107.1 | - | 2 | 748 | |||
AK016438.1 | - | 3 | 1042 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | NM_028688.1 | - | 748 | Chr. - - [-] on Build |