Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 1700029M20Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015613.1 | 3 | 1990 | |||
NR_027909.1 | 2 | 1861 | ||||
GenBank mRNA | AK006510.2 | - | 3 | 1875 | ||
AK016438.1 | - | 3 | 1991 |