Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 1810064F22Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027981.1 | 2 | 1448 | |||
GenBank mRNA | AK142481.1 | - | 2 | 1449 | ||
BC147375.1 | - | 1 | 907 | 9056430 |