Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 1810064F22Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027981.1 | 2 | 394 | |||
GenBank mRNA | AK142481.1 | - | 2 | 395 |