Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc13a2os | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003282.2 | 5 | 1019 | |||
GenBank mRNA | AK040637.1 | - | 5 | 1021 | ||
AK041943.1 | - | 5 | 1021 |