Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Snhg8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_028574.1 | 1 | 725 | |||
GenBank mRNA | AK008369.1 | - | 1 | 176 | ||
BC025067.1 | - | 2 | 471 | |||
BC096391.1 | - | 3 | 313 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BF181273.1 | - | 2 | 470 | Chr. 3 - 123507550 - 123508320 [-] on Build GRCm38 |