Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sprr2j-ps | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003185.1 | 2 | 387 | |||
GenBank mRNA | AK019074.1 | - | 2 | 389 | ||
BC125542.1 | - | 2 | 363 | |||
BC125544.1 | - | 2 | 363 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY708877.1 | - | 2 | 389 | Chr. 3 - 92418087 - 92419398 [+] on Build GRCm38 |