Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wdr73 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027976.1 | 1 | 325 | |||
GenBank mRNA | AK004708.1 | - | 1 | 290 | ||
AK169377.1 | - | 1 | 289 | |||
BC068122.1 | - | 1 | 279 |