Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: B930059L03Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033340.1 | 1 | 852 | |||
GenBank mRNA | AK047421.1 | - | 1 | 853 | ||
AK083693.1 | - | 1 | 467 |
Gene Symbol: 9430024F10Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BB448693.2 | - | 1 | 206 |