Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trub2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110962.1 | 3, 4 | 375 | |||
GenBank mRNA | AK050587.1 | - | 3, 4 | 365 | ||
AK081579.1 | - | 3, 4 | 365 | |||
AU079587.1 | - | 3, 4 | 375 | |||
BU707805.1 | - | 3, 4 | 337 |