Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ythdf3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027375.1 | 3 | 392 | |||
GenBank mRNA | AK039176.1 | - | 3 | 339 | ||
AK053736.1 | - | 3 | 311 | |||
BC057158.1 | - | 3 | 303 | 5348717 |