Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: E130114P18Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015513.2 | 3 | 774 | |||
GenBank mRNA | AK053617.1 | - | 934 | |||
AK082601.1 | - | 3 | 776 | |||
AK086199.1 | - | 1110 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | NM_177023.1 | - | 1110 | Chr. - - [-] on Build |