Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: E130114P18Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015513.2 | 3 | 479 | |||
GenBank mRNA | AK053617.1 | - | 639 | |||
AK082601.1 | - | 3 | 481 | |||
AK086199.1 | - | 815 | ||||
AK086602.1 | - | 3 | 525 | |||
BC048947.1 | - | 4 | 598 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | NM_177023.1 | - | 815 | Chr. - - [-] on Build |