Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gm6548 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003363.1 | 1 | 50 | |||
GenBank mRNA | BC089461.1 | - | 1 | 50 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | CF750205.1 | - | 1 | 88 | Chr. 17 - 78851780 - 78852544 [-] on Build GRCm38 |