Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C130026L21Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015546.2 | 4 | 1708 | |||
GenBank mRNA | AK047985.1 | - | 1550 | |||
AK160721.1 | - | 4 | 1709 | |||
BC094571.1 | - | 3 | 1385 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | NM_177875.1 | - | 1550 | Chr. - - [-] on Build |