Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rhno1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027359.1 | 2 | 146 | |||
NR_027360.1 | 2 | 542 | ||||
GenBank mRNA | AK011109.1 | - | 2 | 237 | ||
AK011167.1 | - | 2 | 340 | |||
AK020031.1 | - | 2 | 395 | |||
AK029454.1 | - | 2 | 228 | |||
AK146167.1 | - | 2 | 117 | |||
AK156717.1 | - | 3 | 660 | |||
AK168313.1 | - | 589 | ||||
BC027368.1 | - | 2 | 544 | |||
BC089484.1 | - | 1 | 94 | |||
BE655013.1 | - | 2 | 146 |
Gene Symbol: Tmlhe | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK051339.1 | - | 3 | 1134 |