Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: D130009I18Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015593.2 | 2 | 326 | |||
GenBank mRNA | AK051169.1 | - | 2 | 330 | ||
AK051495.1 | - | 2 | 326 | |||
AK051771.1 | - | 2 | 331 |