Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sycp1-ps1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024208.1 | 1 | 393 | |||
GenBank mRNA | AK015899.1 | - | 1 | 394 | ||
L41536.1 | - | 1 | 54 |