Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BC064078 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015455.1 | 3 | 406 | |||
GenBank mRNA | AK134020.1 | - | 369 | |||
BC016205.1 | - | 3 | 419 | |||
BC017132.1 | - | 3 | 419 | |||
BC025095.1 | - | 3 | 403 | |||
BC038635.1 | - | 3 | 402 | |||
BC051485.1 | - | 3 | 465 | |||
BC064078.1 | - | 3 | 395 | |||
BC096405.1 | - | 3 | 406 |
Gene Symbol: B230317F23Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK045882.1 | - | 1 | 1191 | ||
AK050074.1 | - | 1 | 1170 |