Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gt(ROSA)26Sor | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
RefSeq | NR_027008.1 | 2 | 206 | |||
NR_027009.1 | 2 | 100 | ||||
NR_027010.1 | 2 | 100 | ||||
GenBank mRNA | AK021028.1 | - | 2 | 47 | ||
BY207362.1 | - | 2 | 101 | |||
BY727583.1 | - | 2 | 45 | |||
U83174.1 | - | LIT3416 Rosa26.1 |
2 | 159 | ||
U83175.1 | - | LIT3417 Rosa26.2 |
2 | 139 |