Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gorasp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027343.1 | 1, 2 | 261 | |||
GenBank mRNA | AK002515.1 | - | 1, 2 | 143 | ||
AK168928.1 | - | 1, 2 | 194 | |||
AK210678.1 | - | 1 | 19 |