Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gorasp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027343.1 | 2 | 403 | |||
GenBank mRNA | AK002515.1 | - | 2 | 285 | ||
AK168928.1 | - | 2 | 336 | |||
AK210678.1 | - | 1 | 161 |