Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 5330426P16Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_028300.1 | 2 | 657 | |||
GenBank mRNA | AK019912.1 | - | 568 | |||
AK035047.1 | - | 613 | ||||
AK045752.1 | - | 2 | 622 | |||
AK052569.1 | - | 2 | 628 | |||
AK137010.1 | - | 2 | 1108 | |||
AK151508.1 | - | 634 |