Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 5330426P16Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_028300.1 | 2 | 1697 | |||
GenBank mRNA | AK035047.1 | - | 1653 | |||
AK137010.1 | - | 2 | 2148 |