Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pram1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_028279.1 | 1 | 84 | |||
GenBank mRNA | AK169990.1 | - | 1 | 78 | ||
BY207392.1 | - | 1 | 85 |