Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 0610012G03Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027897.1 | 1 | 495 | |||
GenBank mRNA | AK002600.1 | - | 1 | 466 | ||
AK002881.1 | - | 1 | 431 | |||
AK009191.1 | - | 1 | 473 | |||
AK011122.1 | - | 1 | 460 | |||
AK146135.1 | - | 1 | 487 | |||
BC021536.1 | - | 1 | 465 |