Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 4933407K13Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_029443.1 | 9 | 1346 | |||
GenBank mRNA | AK030939.1 | - | 10 | 1347 | ||
AK036260.1 | - | 1 | 1266 | |||
AK053637.1 | - | 9 | 1460 | |||
AK163097.1 | - | 1346 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB617014.1 | - | 2 | 259 | Chr. X - 75727098 - 75746904 [-] on Build GRCm38 |