Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 1810032O08Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027819.1 | 1 | 79 | |||
NR_027820.1 | 1 | 79 | ||||
NR_027821.1 | 1 | 79 | ||||
GenBank mRNA | AK007675.1 | - | 1 | 80 | ||
AK013681.1 | - | 1 | 71 | |||
AK037979.1 | - | 1 | 71 | |||
AK088915.1 | - | 1 | 59 | |||
BC055707.1 | - | 1 | 65 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | NM_025472.1 | - | 71 | Chr. - - [-] on Build |