Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 5430405H02Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015591.1 | 1 | 74 | |||
NR_027914.1 | 1 | 74 | ||||
GenBank mRNA | AK012005.1 | - | 1 | 81 | ||
AK017271.1 | - | 93 | ||||
AK134903.1 | - | 1 | 76 | |||
AK134962.1 | - | 1 | 24 |