Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 5430405H02Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015591.1 | 2 | 932 | |||
NR_027914.1 | 2 | 646 | ||||
GenBank mRNA | AK134903.1 | - | 2 | 648 | ||
AK134962.1 | - | 2 | 882 | |||
BC042697.1 | - | 2 | 753 |