Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kctd18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027630.1 | 3, 4 | 531 | |||
GenBank mRNA | BC075692.1 | - | 3, 4 | 531 | 30631161 |