Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 2010204K13Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027924.1 | 2 | 185 | |||
NR_027925.1 | 2 | 195 | ||||
GenBank mRNA | AF038507.1 | - | 2 | 149 | ||
AK002502.1 | - | 534 | ||||
AK008437.1 | - | 2 | 186 | |||
AK077356.1 | - | 2 | 229 | |||
BC042200.1 | - | 2 | 148 | |||
BC061056.1 | - | 2 | 196 | |||
BC072610.1 | - | 2 | 109 |